Agar, powder

Supplier: Thermo Scientific Chemicals

A10752-0E A10752-22 A10752-36
CAAAA10752-0EEA 921.14 CAD
CAAAA10752-0E CAAAA10752-22 CAAAA10752-36
Agar, powder
Agar

MDL: MFCD00081288


CAS No.: 9002-18-0 Molecular Formula: (C12H18O9)X Physical Form: Powder Appearance: White to cream Odor: Odorless Merck Reference: 14,184 MDL No.: MFCD00081288

Melting Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081288
CAS Number: 9002-18-0
EINECS: 232-658-1
Merck Index: 14,184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR