2,3,5-Trimethylphenol 98+%

Supplier: Thermo Scientific Chemicals
Danger

A10745-14 A10745-22 A10745-0B
CAAAAA10745-14EA 45.98 CAD
CAAAAA10745-14 CAAAAA10745-22 CAAAA10745-0B
2,3,5-Trimethylphenol 98+%
2,3,5-Trimethylphenol

MDL: MFCD00002228 Beilstein Registry No.: 2042210


Purity: 98+% CAS No.: 697-82-5 Molecular Formula: C9H12O Formula Weight: 136.20 Odor: Phenol-like Melting Point: 92-95° Boiling Point: 230-231° Beilstein Reference: 2042210 UN No.: 2430 MDL No.: MFCD00002228

Formula: C₉H₁₂O
Boiling Pt: 230…231 °C
Melting Pt: 92…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00002228
CAS Number: 697-82-5
EINECS: 211-806-9
UN: 2430
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR