Heptafluorobutyric anhydride (HFAA), anhydrous 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10744-06
A10744-14
A10744-22
CAAAA10744-06EA
84.37
CAD
CAAAA10744-06
CAAAA10744-14
CAAAA10744-22
Heptafluorobutyric anhydride (HFAA), anhydrous 98%
Heptafluorobutyric anhydride
MDL: MFCD00000432
Purity: 98%
CAS No.: 336-59-4
Molecular Formula: C8F14O3
Formula Weight: 410.06
Refractive Index: 1.2870
Odor: Stench
Boiling Point: 108-110°
UN No.: 3265
MDL No.: MFCD00000432
Formula:
C₈F₁₄O₃ Boiling Pt: 108…110 °C Melting Pt: –43 °C Density: 1.662 Storage Temperature: Ambient |
MDL Number:
MFCD00000432 CAS Number: 336-59-4 EINECS: 206-410-8 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...