2-Chloro-4-iodoaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10742-22
A10742-06
A10742-14
CAAAAA10742-22EA
842.14
CAD
CAAAAA10742-22
CAAAA10742-06
CAAAA10742-14
2-Chloro-4-iodoaniline 98%
2-Chloro-4-iodoaniline
MDL: MFCD00025298
Beilstein Registry No.: 2081106
Purity: 98%
CAS No.: 42016-93-3
Molecular Formula: C6H5ClIN
Formula Weight: 253.47
Melting Point: 68-72°
Beilstein Reference: 2081106
UN No.: 2811
MDL No.: MFCD00025298
Formula:
C₆H₅ClIN Boiling Pt: 289 °C (1013 hPa) Melting Pt: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00025298 CAS Number: 42016-93-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...