Maltol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10735-14
A10735-22
CAAAA10735-14EA
58.3
CAD
CAAAA10735-14
CAAAA10735-22
Maltol 99%
Maltol
MDL: MFCD00006578
Beilstein Registry No.: 112169
Purity: 99%
CAS No.: 118-71-8
Molecular Formula: C6H6O3
Formula Weight: 126.11
Flash Point: 95°(203°F)
Melting Point: 161-165°
Boiling Point: 170°/100mm
Beilstein Reference: 112169
Merck Reference: 14,5713
MDL No.: MFCD00006578
Formula:
C₆H₆O₃ Boiling Pt: 170 °C/100 mm Melting Pt: 161…165 °C Density: 1.453 g/cm³ (20 °C) Flash Pt: 95 °C(203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006578 CAS Number: 118-71-8 EINECS: 204-271-8 Merck Index: 14,5713 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...