4-Iodobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Iodobenzoic acid

A10730-09 A10730-18 A10730-30
CAAAA10730-09EA 67.62 CAD
CAAAA10730-09 CAAAA10730-18 CAAAA10730-30
4-Iodobenzoic acid 97%
4-Iodobenzoic acid

MDL: MFCD00002533 Beilstein Registry No.: 1860232


Purity: 97% CAS No.: 619-58-9 Molecular Formula: C7H5IO2 Formula Weight: 248.02 Melting Point: ca 265° dec. Beilstein Reference: 1860232

Formula: C₇H₅IO₂
MW: 248.02 g/mol
Boiling Pt: 319
Melting Pt: ∼ 265 °C (dec.)
Density: 2.18
Storage Temperature: Ambient
MDL Number: MFCD00002533
CAS Number: 619-58-9
EINECS: 210-603-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR