3-Hydroxy-4-nitrobenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10724-06
A10724-14
A10724-22
CAAAA10724-06EA
49.93
CAD
CAAAA10724-06
CAAAA10724-14
CAAAA10724-22
3-Hydroxy-4-nitrobenzoic acid 98%
3-Hydroxy-4-nitrobenzoic acid
MDL: MFCD00007110
Beilstein Registry No.: 2107564
Purity: 98%
CAS No.: 619-14-7
Molecular Formula: C7H5NO5
Formula Weight: 183.12
Melting Point: 232-236°
Beilstein Reference: 2107564
Formula:
C₇H₅NO₅ Melting Pt: 232…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007110 CAS Number: 619-14-7 EINECS: 210-580-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...