4-Bromo-3-nitrotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10718-14
A10718-22
CAAAA10718-14EA
162.74
CAD
CAAAA10718-14
CAAAA10718-22
4-Bromo-3-nitrotoluene 99%
4-Bromo-3-nitrotoluene
MDL: MFCD00024180
Beilstein Registry No.: 1945925
Purity: 98+%
CAS No.: 5326-34-1
Molecular Formula: C7H6BrNO2
Formula Weight: 216.04
Flash Point:
Formula:
C₇H₆BrNO₂ Boiling Pt: 96 °C/1 mm Melting Pt: 29…32 °C Density: 1.578 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00024180 CAS Number: 5326-34-1 EINECS: 226-203-6 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...