4-Bromo-2,6-xylidine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Bromo-2,6-dimethylaniline

A10712-14 A10712-30 A10712-09 A10712-18
CAAAAA10712-14EA 129.72 CAD
CAAAAA10712-14 CAAAAA10712-30 CAAAA10712-09 CAAAA10712-18
4-Bromo-2,6-xylidine 98%
4-Bromo-2,6-xylidine

MDL: MFCD00007826 Beilstein Registry No.: 1936300


Purity: 98% CAS No.: 24596-19-8 Molecular Formula: C8H10BrN Formula Weight: 200.08 Flash Point: 110°(230°F) Melting Point: 47-52° Boiling Point: 120°/0.2mm Beilstein Reference: 1936300 UN No.: 2811 MDL No.: MFCD00007826

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 120 °C/0.2 mm
Melting Pt: 47…52 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007826
CAS Number: 24596-19-8
EINECS: 246-337-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR