2-Chloro-5-nitropyridine 99%

Supplier: Thermo Scientific Chemicals
Warning

A10709-14 A10709-22
CAAAA10709-14EA 83.74 CAD
CAAAA10709-14 CAAAA10709-22
2-Chloro-5-nitropyridine 99%
2-Chloro-5-nitropyridine

MDL: MFCD00006240 Beilstein Registry No.: 120453


Purity: 99% CAS No.: 4548-45-2 Molecular Formula: C5H3ClN2O2 Formula Weight: 158.54 Flash Point: 158°(316°F) Melting Point: 104-109° Beilstein Reference: 120453 MDL No.: MFCD00006240

Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 104…109 °C
Flash Pt: 158 °C(316 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006240
CAS Number: 4548-45-2
EINECS: 224-908-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR