Ammonium formate, (max. 3% H₂O) 97% (by acidometric titration)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Formic acid ammonium salt

A10699-0B A10699-0I A10699-30
CAAAA10699-0BEA 167.48 CAD
CAAAA10699-0B CAAAA10699-0I CAAAA10699-30
Ammonium formate, (max. 3% H₂O) 97% (by acidometric titration)
Ammonium formate

MDL: MFCD00013103 Beilstein Registry No.: 3625095 Fieser: 1,38 15,13 18,23 20,16


Purity: 97% CAS No.: 540-69-2 Molecular Formula: CH5NO2 Formula Weight: 63.06 Odor: Ammonia-like Melting Point: ca 116° Beilstein Reference: 3625095 Merck Reference: 14,523 MDL No.: MFCD00013103

Formula: HCO₂NH₄
Melting Pt: ∼ 116 °C
Density: 1.28
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Merck Index: 14,523

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR