2-Adamantanol 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
2-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-2-ol, 2-Tricyclo[3.3.1.1(3,7)]decanol
A10697-06
A10697-14
CAAAA10697-06EA
73.63
CAD
CAAAA10697-06
CAAAA10697-14
2-Adamantanol 98%
2-Adamantanol
MDL: MFCD00074744
Beilstein Registry No.: 1903957
Purity: 98%
CAS No.: 700-57-2
Molecular Formula: C10H16O
Formula Weight: 152.24
Melting Point: 299-303°(sealed tube)
Beilstein Reference: 1903957
Formula:
C₁₀H₁₆O Melting Pt: 299…303 °C (sealed tube) Density: 1.07 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00074744 CAS Number: 700-57-2 EINECS: 211-846-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...