1-Bromobutane 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Butyl bromide
A10696-0C
A10696-0E
A10696-36
CAAAA10696-0CEA
665.18
CAD
CAAAA10696-0C
CAAAA10696-0E
CAAAA10696-36
1-Bromobutane 98+%
1-Bromobutane
MDL: MFCD00000260
Beilstein Registry No.: 1098260
Purity: 98+%
CAS No.: 109-65-9
Molecular Formula: C4H9Br
Formula Weight: 137.03
Flash Point: 20°(68°F)
Physical Form: Liquid
Density: 1.275
Refractive Index: 1.4400
Appearance: Colorless
Odor: Characteristic
Melting Point: -112°
Boiling Point: 101-102°
Beilstein Reference: 1098260
Merck Reference: 14,1553
UN No.: 1126
MDL No.: MFCD00000260
Formula:
BrH₂C(CH₂)₂CH₃ Boiling Pt: 101…102 °C Melting Pt: –112 °C Density: 1.275 Flash Pt: 20 °C (68 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000260 CAS Number: 109-65-9 EINECS: 203-691-9 UN: 1126 ADR: 3,II Merck Index: 14,1553 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...