4-Fluoro-2-nitroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10685-18
A10685-36
A10685-22
CAAAAA10685-18EA
102.38
CAD
CAAAAA10685-18
CAAAAA10685-36
CAAAA10685-22
4-Fluoro-2-nitroaniline 98%
4-Fluoro-2-nitroaniline
MDL: MFCD00007830
Beilstein Registry No.: 2210197
Purity: 98%
CAS No.: 364-78-3
Molecular Formula: C6H5FN2O2
Formula Weight: 156.12
Flash Point: 89°(192°F)
Melting Point: 90-94°
Beilstein Reference: 2210197
MDL No.: MFCD00007830
Formula:
C₆H₅FN₂O₂ Melting Pt: 90…94 °C Flash Pt: 89 °C(192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007830 CAS Number: 364-78-3 EINECS: 206-666-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...