D-(+)-Proline 99%

Supplier: Thermo Scientific Chemicals

Synonyms: H-D-Pro-OH, (R)-(+)-Proline, D-Proline

A10682-03 A10682-06 A10682-14
CAAAA10682-03EA 79 CAD
CAAAA10682-03 CAAAA10682-06 CAAAA10682-14
D-(+)-Proline 99%
D(+)-Proline

MDL: MFCD00064317 Beilstein Registry No.: 80811 Fieser: 7,307 9,393 Optical Rotation: +84°C (c=1 in water)


Purity: 99% CAS No.: 344-25-2 Molecular Formula: C5H9NO2 Formula Weight: 115.13 Melting Point: 213-215° dec. Beilstein Reference: 80811

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 213…215 °C (dec.)
Density: 1.36
Storage Temperature: Ambient
MDL Number: MFCD00064317
CAS Number: 344-25-2
EINECS: 206-452-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR