4-(Trifluoromethoxy)benzaldehyde 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10671-06
A10671-14
CAAAA10671-06EA
75.05
CAD
CAAAA10671-06
CAAAA10671-14
4-(Trifluoromethoxy)benzaldehyde 96%
4-(Trifluoromethoxy)benzaldehyde
MDL: MFCD00041530
Beilstein Registry No.: 1949135
Purity: 96%
CAS No.: 659-28-9
Molecular Formula: C8H5F3O2
Formula Weight: 190.12
Flash Point: 71°(159°F)
Refractive Index: 1.4580
Boiling Point: 93°/27mm
Beilstein Reference: 1949135
MDL No.: MFCD00041530
Formula:
C₈H₅F₃O₂ Boiling Pt: 93 °C/27 mm Density: 1.331 Flash Pt: 71 °C(159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041530 CAS Number: 659-28-9 EINECS: 211-531-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...