(3-Aminopropyl)triethoxysilane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10668-0E
A10668-22
A10668-36
CAAAA10668-0EEA
826.34
CAD
CAAAA10668-0E
CAAAA10668-22
CAAAA10668-36
(3-Aminopropyl)triethoxysilane 98%
(3-Aminopropyl)triethoxysilane
MDL: MFCD00008207
Beilstein Registry No.: 1754988
Purity: 98%
CAS No.: 919-30-2
Molecular Formula: C9H23NO3Si
Formula Weight: 221.37
Flash Point: 104°(219°F)
Refractive Index: 1.4210
Odor: Amine-like
pH: 11
Boiling Point: 217°
Beilstein Reference: 1754988
UN No.: 2735
MDL No.: MFCD00008207
Formula:
H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃ Boiling Pt: 217 °C Melting Pt: < –70 °C Density: 0.948 Flash Pt: 104 °C (219 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008207 CAS Number: 919-30-2 EINECS: 213-048-4 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...