2-Amino-3-hydroxypyridin 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Aminopyridin-3-ol, 2-Amino-3-hydroxypyridine

A10665-09 A10665-18 A10665-30
CAAAA10665-09EA 53.88 CAD
CAAAA10665-09 CAAAA10665-18 CAAAA10665-30
2-Amino-3-hydroxypyridin 98%
2-Amino-3-hydroxypyridin

MDL: MFCD00006317 Beilstein Registry No.: 109868


Purity: 98% CAS No.: 16867-03-1 Molecular Formula: C5H6N2O Formula Weight: 110.12 Odor: Odorless Melting Point: 166-174° Beilstein Reference: 109868 UN No.: 2811 MDL No.: MFCD00006317

Formula: C₅H₆N₂O
MW: 110.12
Boiling Pt: 250
Melting Pt: 166…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00006317
CAS Number: 16867-03-1
EINECS: 240-886-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR