2-Benzylpyridine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10662-0B
A10662-18
A10662-30
CAAAA10662-0BEA
472.42
CAD
CAAAA10662-0B
CAAAA10662-18
CAAAA10662-30
2-Benzylpyridine 98+%
2-Benzylpyridine
MDL: MFCD00006352
Beilstein Registry No.: 115875
Purity: 98+%
CAS No.: 101-82-6
Molecular Formula: C12H11N
Formula Weight: 169.23
Flash Point: 125°(257°F)
Refractive Index: 1.5790
Melting Point: 10-12°
Boiling Point: 140-143°/11mm
Beilstein Reference: 115875
MDL No.: MFCD00006352
Formula:
C₁₂H₁₁N MW: 169.23 g/mol Boiling Pt: 140…143 °C/11 mm Melting Pt: 10…12 °C Density: 1.054 Flash Pt: 125 °C(257 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006352 CAS Number: 101-82-6 EINECS: 202-979-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...