α-Bromopropiophenone 96%

Supplier: Thermo Scientific Chemicals
Warning

A10661-14 A10661-22 A10661-36
CAAAA10661-14EA 113.13 CAD
CAAAA10661-14 CAAAA10661-22 CAAAA10661-36
α-Bromopropiophenone 96%
α-Bromopropiophenone

MDL: MFCD00000145 Beilstein Registry No.: 508550


Purity: 96% CAS No.: 2114-00-3 Molecular Formula: C9H9BrO Formula Weight: 213.08 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5720 Odor: Pungent Boiling Point: 245-250° Beilstein Reference: 508550 UN No.: 2927 MDL No.: MFCD00000145

Formula: C₆H₅C(O)CHBrCH₃
Boiling Pt: 245…250 °C
Density: 1.417
Flash Pt: >110 °C (230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000145
CAS Number: 2114-00-3
EINECS: 218-307-5
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR