Ethyl decanoate 99%
Supplier: Thermo Scientific Chemicals
A10657-0E
A10657-22
A10657-36
CAAAA10657-0EEA
685.72
CAD
CAAAA10657-0E
CAAAA10657-22
CAAAA10657-36
Ethyl decanoate 99%
Ethyl decanoate
MDL: MFCD00009581
Beilstein Registry No.: 1762128
Purity: 99%
CAS No.: 110-38-3
Molecular Formula: CH3(CH2)8CO2CH2CH3
Formula Weight: 200.32
Flash Point: 102°(216°F)
Refractive Index: 1.4250
Melting Point: -20°
Boiling Point: 250-252°
Beilstein Reference: 1762128
Merck Reference: 14,3776
MDL No.: MFCD00009581
Formula:
C₁₂H₂₄O₂ Boiling Pt: 250…252 °C Melting Pt: –20 °C Density: 0.862 Flash Pt: 102 °C(216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009581 CAS Number: 110-38-3 EINECS: 203-761-9 Merck Index: 14,3776 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...