1-Bromo-3,3-dimethylbutanone 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Bromopinacolone, 1-Bromo-3,3-dimethyl-2-butanone
A10655-09
A10655-18
A10655-30
CAAAA10655-09EA
110.28
CAD
CAAAA10655-09
CAAAA10655-18
CAAAA10655-30
1-Bromo-3,3-dimethylbutanone 97+%
1-Bromo-3,3-dimethylbutanone
MDL: MFCD00000206
Beilstein Registry No.: 506485
Purity: 97+%
CAS No.: 5469-26-1
Molecular Formula: C6H11BrO
Formula Weight: 179.06
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Refractive Index: 1.4655
Odor: Pungent
Melting Point: -10°
Boiling Point: 189-192°
Beilstein Reference: 506485
UN No.: 2810
MDL No.: MFCD00000206
Formula:
C₆H₁₁BrO Boiling Pt: 189…192 °C Melting Pt: –10 °C Density: 1.333 Flash Pt: 75 °C(167 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000206 CAS Number: 5469-26-1 EINECS: 226-794-0 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...