4,4'-Difluorobenzophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(4-Fluorophenyl)methanone, Bis(4-fluorophenyl) ketone

A10653-30 A10653-09 A10653-18
CAAAAA10653-30EA 925.88 CAD
CAAAAA10653-30 CAAAA10653-09 CAAAA10653-18
4,4'-Difluorobenzophenone 98+%
4,4'-Difluorobenzophenone

MDL: MFCD00000353 Beilstein Registry No.: 516231


Purity: 98+% CAS No.: 345-92-6 Molecular Formula: C13H8F2O Formula Weight: 218.20 Melting Point: 106-110° Boiling Point: 170-172°/10mm MDL No.: MFCD00000353

Formula: C₁₃H₈F₂O
Boiling Pt: 170…172 °C/10 mm
Melting Pt: 106…110 °C
Density: 1,39 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000353
CAS Number: 345-92-6
EINECS: 206-466-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR