2,5-Difluorobenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10651-06
A10651-14
CAAAA10651-06EA
76.16
CAD
CAAAA10651-06
CAAAA10651-14
2,5-Difluorobenzoyl chloride 98%
2,5-Difluorobenzoyl chloride
MDL: MFCD00009929
Beilstein Registry No.: 2046666
Purity: 98%
CAS No.: 35730-09-7
Molecular Formula: C7H3ClF2O
Formula Weight: 176.55
Flash Point: 93°(199°F)
Refractive Index: 1.5150
Boiling Point: 81-83°/14mm
Beilstein Reference: 2046666
UN No.: 3265
MDL No.: MFCD00009929
Formula:
C₇H₃ClF₂O Boiling Pt: 81…83 °C/14 mm Density: 1.425 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009929 CAS Number: 35730-09-7 EINECS: 252-701-8 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...