(4-Fluorophenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A10646-06 A10646-14 A10646-22
CAAAA10646-06EA 55.62 CAD
CAAAA10646-06 CAAAA10646-14 CAAAA10646-22
(4-Fluorophenyl)acetic acid 98%
(4-Fluorophenyl)acetic acid

MDL: MFCD00004343 Beilstein Registry No.: 972145


Purity: 98% CAS No.: 405-50-5 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Flash Point: 100°(212°F) Odor: Odorless Boiling Point: 164°/2mm Beilstein Reference: 972145 Merck Reference: 14,4177 MDL No.: MFCD00004343

Formula: (C₆H₄F)CH₂COOH
Boiling Pt: 164 °C/2 mm
Melting Pt: 81…85 °C
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004343
CAS Number: 405-50-5
EINECS: 206-972-4
Merck Index: 14,4177

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR