3-Fluoro-o-xylene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10637-22
A10637-06
A10637-14
CAAAAA10637-22EA
491.38
CAD
CAAAAA10637-22
CAAAA10637-06
CAAAA10637-14
3-Fluoro-o-xylene 99%
3-Fluoro-o-xylene
MDL: MFCD00000323
Beilstein Registry No.: 2040955
Purity: 99%
CAS No.: 443-82-3
Molecular Formula: C8H9F
Formula Weight: 124.16
Flash Point: 36°(96°F)
Refractive Index: 1.4860
Odor: Aromatic
Boiling Point: 147-148°
Beilstein Reference: 2040955
UN No.: 1993
MDL No.: MFCD00000323
Formula:
C₈H₉F Boiling Pt: 147…148 °C Density: 0.992 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000323 CAS Number: 443-82-3 EINECS: 207-140-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...