1-Bromo-2-fluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10635-14
A10635-22
A10635-36
CAAAA10635-14EA
93.69
CAD
CAAAA10635-14
CAAAA10635-22
CAAAA10635-36
1-Bromo-2-fluorobenzene 99%
1-Bromo-2-fluorobenzene
MDL: MFCD00000282
Beilstein Registry No.: 1071819
Fieser: 1,77
Purity: 99%
CAS No.: 1072-85-1
Molecular Formula: C6H4BrF
Formula Weight: 175.01
Flash Point: 43°(109°F)
Refractive Index: 1.5340
Odor: Aromatic
Melting Point: -8°
Boiling Point: 155-157°
Beilstein Reference: 1071819
UN No.: 1993
MDL No.: MFCD00000282
Formula:
C₆H₄BrF Boiling Pt: 155…157 °C Melting Pt: –8 °C Density: 1.601 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000282 CAS Number: 1072-85-1 EINECS: 214-018-3 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...