6-Nitrobenzothiazole 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10634-06
A10634-18
CAAAAA10634-06EA
68.89
CAD
CAAAAA10634-06
CAAAA10634-18
6-Nitrobenzothiazole 98+%
6-Nitrobenzothiazole
MDL: MFCD00014569
Beilstein Registry No.: 7824
Purity: 98+%
CAS No.: 2942-06-5
Molecular Formula: C7H4N2O2S
Formula Weight: 180.18
Melting Point: 173-177°
Beilstein Reference: 7824
Formula:
C₇H₄N₂O₂S Melting Pt: 173…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014569 CAS Number: 2942-06-5 EINECS: 220-933-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...