1-Bromo-2,6-difluorobenzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10633-06
A10633-14
CAAAA10633-06EA
77.89
CAD
CAAAA10633-06
CAAAA10633-14
1-Bromo-2,6-difluorobenzene 98%
1-Bromo-2,6-difluorobenzene
MDL: MFCD00009894
Beilstein Registry No.: 1681053
Purity: 98%
CAS No.: 64248-56-2
Molecular Formula: C6H3BrF2
Formula Weight: 192.99
Flash Point: 53°(127°F)
Refractive Index: 1.5110
Boiling Point: 61°/35mm
Beilstein Reference: 1681053
UN No.: 1993
MDL No.: MFCD00009894
Formula:
C₆H₃BrF₂ Boiling Pt: 61 °C/35 mm Density: 1.710 Flash Pt: 53 °C (127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009894 CAS Number: 64248-56-2 EINECS: 264-750-2 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...