Ethyl-3-nitrobenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10626-14
A10626-22
CAAAA10626-14EA
94.96
CAD
CAAAA10626-14
CAAAA10626-22
Ethyl-3-nitrobenzoate 98+%
Ethyl 3-nitrobenzoate
MDL: MFCD00014702
Beilstein Registry No.: 1912880
Purity: 98+%
CAS No.: 618-98-4
Molecular Formula: C9H9NO4
Formula Weight: 195.18
Melting Point: 40-43°
Boiling Point: 297-298°
Merck Reference: 14,3832
MDL No.: MFCD00014702
Formula:
C₉H₉NO₄ Boiling Pt: 297…298 °C Melting Pt: 40…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014702 CAS Number: 618-98-4 EINECS: 210-574-6 Merck Index: 14,3832 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...