1-Nitroso-2-naphthol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10622-09
A10622-18
A10622-30
CAAAA10622-09EA
54.19
CAD
CAAAA10622-09
CAAAA10622-18
CAAAA10622-30
1-Nitroso-2-naphthol 99%
1-Nitroso-2-naphthol
MDL: MFCD00003884
Beilstein Registry No.: 776947
Purity: 99%
CAS No.: 131-91-9
Molecular Formula: C10H7NO2
Formula Weight: 173.17
Melting Point: 104-108° dec.
Beilstein Reference: 776947
Merck Reference: 14,6641
UN No.: 3077
MDL No.: MFCD00003884
Formula:
C₁₀H₇NO₂ Boiling Pt: 304 °C (1013 hPa) Melting Pt: 104…108 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003884 CAS Number: 131-91-9 EINECS: 205-043-0 UN: 3077 ADR: 9,III Merck Index: 14,6641 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...