Dimethyl-5-nitroisophthalate 98+%

Supplier: Thermo Scientific Chemicals

A10610-0B A10610-30
CAAAA10610-0BEA 363.4 CAD
CAAAA10610-0B CAAAA10610-18 CAAAA10610-30
Dimethyl-5-nitroisophthalate 98+%
Dimethyl 5-nitroisophthalate

MDL: MFCD00008429 Beilstein Registry No.: 2140916


Purity: 98+% CAS No.: 13290-96-5 Molecular Formula: C10H9NO6 Formula Weight: 239.19 Melting Point: 121-123° Beilstein Reference: 2140916

Formula: C₁₀H₉NO₆
Melting Pt: 121…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00008429
CAS Number: 13290-96-5
EINECS: 236-307-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR