2,3,4-Trimethoxybenzaldehyde 98+%

Supplier: Thermo Scientific Chemicals

A10609-09 A10609-18 A10609-30
CAAAA10609-09EA 48.35 CAD
CAAAA10609-09 CAAAA10609-18 CAAAA10609-30
2,3,4-Trimethoxybenzaldehyde 98+%
2,3,4-Trimethoxybenzaldehyde

MDL: MFCD00003310 Beilstein Registry No.: 981091


Purity: 98+% CAS No.: 2103-57-3 Molecular Formula: C10H12O4 Formula Weight: 196.20 Flash Point: 110°(230°F) Refractive Index: 1.5547 Melting Point: 38-42° Boiling Point: 168-170°/12mm Beilstein Reference: 981091 MDL No.: MFCD00003310

Formula: C₁₀H₁₂O₄
MW: 196.2 g/mol
Boiling Pt: 168…170 °C/12 mm
Melting Pt: 38…42 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003310
CAS Number: 2103-57-3
EINECS: 218-271-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR