Methyl bromoacetate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Bromoacetic methyl ester
A10605-0B
A10605-18
CAAAA10605-0BEA
581.44
CAD
CAAAA10605-0B
CAAAA10605-18
Methyl bromoacetate 98+%
Methyl bromoacetate
MDL: MFCD00000189
Beilstein Registry No.: 506256
Purity: 98+%
CAS No.: 96-32-2
Molecular Formula: C3H5BrO2
Formula Weight: 152.98
Flash Point: 62°(143°F)
Refractive Index: 1.4585
Odor: Pungent
Melting Point: -50°
Boiling Point: 144-145°
Beilstein Reference: 506256
UN No.: 2643
MDL No.: MFCD00000189
Formula:
BrCH₂COOCH₃ MW: 152.98 g/mol Boiling Pt: 145 Melting Pt: -50 Density: 1.655 Flash Pt: 62 °C (143 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000189 CAS Number: 96-32-2 EINECS: 202-499-2 UN: 2643 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...