3-Bromo-2,5-dichlorothiophene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10603-03
A10603-06
A10603-14
CAAAA10603-03EA
45.19
CAD
CAAAA10603-03
CAAAA10603-06
CAAAA10603-14
3-Bromo-2,5-dichlorothiophene 97%
3-Bromo-2,5-dichlorothiophene
MDL: MFCD00041440
Beilstein Registry No.: 1306272
Purity: 97%
CAS No.: 60404-18-4
Molecular Formula: C4HBrCl2S
Formula Weight: 231.93
Refractive Index: 1.6080
Boiling Point: 95-97°/14mm
MDL No.: MFCD00041440
Formula:
C₄HBrCl₂S Boiling Pt: 95…97 °C/14 mm Storage Temperature: Ambient |
MDL Number:
MFCD00041440 CAS Number: 60404-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...