1-Bromododecane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10600-0B
A10600-0I
A10600-30
CAAAA10600-0BEA
178.54
CAD
CAAAA10600-0B
CAAAA10600-0I
CAAAA10600-30
1-Bromododecane 98%
1-Bromododecane
MDL: MFCD00000225
Beilstein Registry No.: 506159
Purity: 98%
CAS No.: 143-15-7
Molecular Formula: C12H25Br
Formula Weight: 249.24
Flash Point: 146°(294°F)
Refractive Index: 1.4580
Odor: Weak
Melting Point: -10°
Boiling Point: 276°
Beilstein Reference: 506159
Merck Reference: 14,5389
MDL No.: MFCD00000225
Formula:
BrH₂C(CH₂)₁₀CH₃ Boiling Pt: 276 °C Melting Pt: –10 °C Density: 1.040 Flash Pt: 146 °C(294 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000225 CAS Number: 143-15-7 EINECS: 205-587-9 Merck Index: 14,5389 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...