Trimethyl phosphate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10595-0E
A10595-36
CAAAA10595-0EEA
432.92
CAD
CAAAA10595-0E
CAAAA10595-36
Trimethyl phosphate 98+%
Trimethyl phosphate
MDL: MFCD00008348
Beilstein Registry No.: 1071731
Purity: 98+%
CAS No.: 512-56-1
Molecular Formula: C3H9O4P
Formula Weight: 140.08
Flash Point:
Formula:
C₃H₉O₄P Boiling Pt: 196…198 °C Melting Pt: –46 °C Density: 1.213 Flash Pt: >150 °C(302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008348 CAS Number: 512-56-1 EINECS: 208-144-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...