4,4'-Dimethoxybenzil 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione

A10594-06 A10594-14 A10594-22
CAAAA10594-06EA 92.75 CAD
CAAAA10594-06 CAAAA10594-14 CAAAA10594-22
4,4'-Dimethoxybenzil 98+%
4,4'-Dimethoxybenzil

MDL: MFCD00008405 Beilstein Registry No.: 531345


Purity: 98+% CAS No.: 1226-42-2 Molecular Formula: C16H14O4 Formula Weight: 270.29 Melting Point: 132-134° Beilstein Reference: 531345

Formula: C₁₆H₁₄O₄
MW: 270.28 g/mol
Boiling Pt: 215 - 219 °C (3 -rr)
Melting Pt: 132…134 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008405
CAS Number: 1226-42-2
EINECS: 214-960-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR