Biphenyl-2-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenylphenol, 2-Hydroxydiphenyl, 2-Hydroxybiphenyl

A10592-30 A10592-0E A10592-36
CAAAAA10592-30EA 69.2 CAD
CAAAAA10592-30 CAAAA10592-0E CAAAA10592-36
Biphenyl-2-ol 99%
Biphenyl-2-ol

MDL: MFCD00002208 Beilstein Registry No.: 606907


Purity: 99% CAS No.: 90-43-7 Molecular Formula: C12H10O Formula Weight: 170.21 Flash Point: 124°(255°F) Odor: Mild Boiling Point: 152-154°/15mm Beilstein Reference: 606907 Merck Reference: 14,7304 UN No.: 3077 MDL No.: MFCD00002208

Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 152…154 °C (15 mmHg)
Melting Pt: 54…58 °C
Density: 1.293
Flash Pt: 124 °C (255 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002208
CAS Number: 90-43-7
EINECS: 201-993-5
UN: 3077
ADR: 9 III
Merck Index: 14,7304

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR