DL-Leucine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-4-methylpentanoic acid, DL-Leucine, (±)-Leucine

A10590-0B A10590-18 A10590-30
CAAAA10590-0BEA 1027 CAD
CAAAA10590-0B CAAAA10590-18 CAAAA10590-30
DL-Leucine 99%
DL-Leucine

MDL: MFCD00063087 Beilstein Registry No.: 636005


Purity: 99% CAS No.: 328-39-2 Molecular Formula: C6H13NO2 Formula Weight: 131.18 Melting Point: ca 294° subl. Beilstein Reference: 636005 Merck Reference: 14,5451 MDL No.: MFCD00063087

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 226
Melting Pt: ∼ 294 °C (subl.)
Density: 1.293
Storage Temperature: Ambient
MDL Number: MFCD00063087
CAS Number: 328-39-2
EINECS: 206-328-2
Merck Index: 14,5451

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR