2,4-Dimethylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10589-09
A10589-18
CAAAA10589-09EA
79.32
CAD
CAAAA10589-09
CAAAA10589-18
2,4-Dimethylbenzoic acid 98%
2,4-Dimethylbenzoic acid
MDL: MFCD00002480
Beilstein Registry No.: 971589
Purity: 98%
CAS No.: 611-01-8
Molecular Formula: C9H10O2
Formula Weight: 150.18
Flash Point: 154°(309°F)
Odor: Odorless
Boiling Point: 267°/727mm
Beilstein Reference: 971589
MDL No.: MFCD00002480
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Boiling Pt: 267 Melting Pt: 124…126 °C Flash Pt: 154 °C (309 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002480 CAS Number: 611-01-8 EINECS: 210-246-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...