1-Ethylquinaldinium iodide 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Ethyl-2-methylquinolin-1-ium iodide, 1-Ethyl-2-methylquinolinium iodide

A10584-06 A10584-14 A10584-22
CAAAA10584-06EA 101.28 CAD
CAAAA10584-06 CAAAA10584-14 CAAAA10584-22
1-Ethylquinaldinium iodide 97%
1-Ethylquinaldinium iodide

MDL: MFCD00031829 Notes: Precursor in synthesis of carbocyanine dyes.


Purity: 97% CAS No.: 606-55-3 Molecular Formula: C12H14IN Formula Weight: 299.16 Melting Point: 231° dec. MDL No.: MFCD00031829

Formula: C₁₂H₁₄IN
Melting Pt: 231 °C (dec.)
Density: 1.689 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00031829
CAS Number: 606-55-3
EINECS: 210-120-7
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR