Isoniazid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10583-22
A10583-36
A10583-0B
A10583-18
CAAAAA10583-22EA
53.56
CAD
CAAAAA10583-22
CAAAAA10583-36
CAAAA10583-0B
CAAAA10583-18
Isoniazid 98+%
Isoniazid
Used for fluorescent HPLC detection of delta 4-3-ketosteroids
Purity: 98+%
CAS No.: 54-85-3
Molecular Formula: C6H7N3O
Formula Weight: 137.14
Melting Point: 169-174°
Beilstein Reference: 119374
Merck Reference: 14,5186
MDL No.: MFCD00006426
Formula:
C₆H₇N₃O Melting Pt: 169…174 °C Density: 1.42 g/cm³ (20 °C) Flash Pt: ≥ 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006426 CAS Number: 54-85-3 EINECS: 200-214-6 Merck Index: 14,5186 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...