3-Nitrobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Nitrobenzoyl chloride

A10579-18 A10579-30
CAAAA10579-0BEA 1086.95 CAD
CAAAA10579-0B CAAAA10579-18 CAAAA10579-30
3-Nitrobenzoyl chloride 98%
3-Nitrobenzoyl chloride

MDL: MFCD00007247 Beilstein Registry No.: 777186


Purity: 98% CAS No.: 121-90-4 Molecular Formula: C7H4ClNO3 Formula Weight: 185.57 Flash Point: 110°(230°F) Odor: Pungent Boiling Point: 276-278° dec. Beilstein Reference: 777186 UN No.: 3261 MDL No.: MFCD00007247

Formula: C₇H₄ClNO₃
MW: 185.57 g/mol
Boiling Pt: 275 (dec.)
Melting Pt: 31…35 °C
Density: 1.42
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007247
CAS Number: 121-90-4
EINECS: 204-505-9
UN: 1759
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR