1-Bromononane 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Nonyl bromide

A10577-14 A10577-22 A10577-36
CAAAA10577-14EA 70.78 CAD
CAAAA10577-14 CAAAA10577-22 CAAAA10577-36
1-Bromononane 99%
1-Bromononane

MDL: MFCD00000278 Beilstein Registry No.: 1733632


Purity: 99% CAS No.: 693-58-3 Molecular Formula: CH3(CH2)8Br Formula Weight: 207.16 Flash Point: 90°(194°F) Refractive Index: 1.4550 Boiling Point: 200-201° Beilstein Reference: 1733632 MDL No.: MFCD00000278

Formula: BrH₂C(CH₂)₇CH₃
MW: 207.15 g/mol
Boiling Pt: 200…201 °C
Melting Pt: –29 °C
Density: 1.086
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000278
CAS Number: 693-58-3
EINECS: 211-755-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR