D-α-Phenylalanine 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
D-Phenylalanine, H-D-Phe-OH, 3-Phenyl-D-alanine, 3-Phenyl-D(+)-alanine
A10572-06
A10572-14
A10572-22
CAAAA10572-06EA
52.77
CAD
CAAAA10572-06
CAAAA10572-14
CAAAA10572-22
D-α-Phenylalanine 99%
D-α-Phenylalanine
MDL: MFCD00004270
Beilstein Registry No.: 2804068
Optical Rotation: +34°C (c=2 in water)
Purity: 99%
CAS No.: 673-06-3
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Melting Point: ca 275° dec.
Beilstein Reference: 2804068
Merck Reference: 14,7271
Optical Rotation: +34° (c=2 in water)
MDL No.: MFCD00004270
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol Boiling Pt: 295 Melting Pt: ∼ 273° … 276 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004270 CAS Number: 673-06-3 EINECS: 211-603-5 Merck Index: 14,7271 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...