3,5-Xylidine 97+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3,5-Dimethylaniline, 5-Amino-m-xylene
A10570-22
A10570-36
CAAAA10570-22EA
104.6
CAD
CAAAA10570-22
CAAAA10570-36
3,5-Xylidine 97+%
3,5-Xylidine
MDL: MFCD00007813
Beilstein Registry No.: 507051
Purity: 97+%
CAS No.: 108-69-0
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 93°(199°F)
Refractive Index: 1.5580
Odor: Characteristic
Melting Point: 9-11°
Boiling Point: 220-221°
Beilstein Reference: 507051
Merck Reference: 14,10084
UN No.: 1711
MDL No.: MFCD00007813
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 220…221 °C Melting Pt: 9…11 °C Density: 0.971 Flash Pt: 93 °C (199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007813 CAS Number: 108-69-0 EINECS: 203-607-0 UN: 1711 ADR: 6.1,II Merck Index: 14,10084 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...