2-Iodobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A10563-14 A10563-22 A10563-36
CAAAA10563-14EA 43.92 CAD
CAAAA10563-14 CAAAA10563-22 CAAAA10563-36
2-Iodobenzoic acid 98+%
2-Iodobenzoic acid

Reagent for detection of sulfhydryl groups in proteins


Purity: 98+% CAS No.: 88-67-5 Molecular Formula: C7H5IO2 Formula Weight: 248.02 Melting Point: 160-164° Beilstein Reference: 1861406 Merck Reference: 14,5030 MDL No.: MFCD00002419

Formula: C₇H₅IO₂
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 160…164 °C
Density: 2.250
Storage Temperature: Ambient
MDL Number: MFCD00002419
CAS Number: 88-67-5
EINECS: 201-850-7
Merck Index: 14,5030

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR