4-Hydroxy-3-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A10562-14 A10562-22
CAAAA10562-14EA 63.04 CAD
CAAAA10562-14 CAAAA10562-22
4-Hydroxy-3-nitrobenzoic acid 98%
4-Hydroxy-3-nitrobenzoic acid

Standard in chymotrypsin assays


Purity: 98% CAS No.: 616-82-0 Molecular Formula: C7H5NO5 Formula Weight: 183.12 Melting Point: 182-184° Beilstein Reference: 2213134

Formula: C₇H₅NO₅
MW: 183.12 g/mol
Melting Pt: 182…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00007119
CAS Number: 616-82-0
EINECS: 210-494-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR