N,N-Diethylaniline 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10560-0D
A10560-0F
A10560-AP
CAAAA10560-0DEA
545.1
CAD
CAAAA10560-0D
CAAAA10560-0F
CAAAA10560-AP
N,N-Diethylaniline 99%
N,N-Diethylaniline
MDL: MFCD00009042
Beilstein Registry No.: 742483
Fieser: 5,212
Purity: 99%
CAS No.: 91-66-7
Molecular Formula: C10H15N
Formula Weight: 149.24
Flash Point: 85°(185°F)
Refractive Index: 1.5420
Melting Point: -38°
Boiling Point: 215-216°
Beilstein Reference: 742483
Merck Reference: 14,3114
UN No.: 2432
MDL No.: MFCD00009042
Formula:
C₁₀H₁₅N MW: 149.24 g/mol Boiling Pt: 215…216 °C Melting Pt: –38 °C Density: 0.936 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009042 CAS Number: 91-66-7 EINECS: 202-088-8 UN: 2432 ADR: 6.1 III Merck Index: 14,3114 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...